missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-4-pyrazolecarbonitrile 98.0+%, TCI America™
Supplier: TCI America A159425G
Specifications
3-Amino-4-pyrazolecarbonitrile | |
175°C | |
C4H4N4 | |
25 g | |
FFNKBQRKZRMYCL-UHFFFAOYSA-N | |
5-amino-1H-pyrazole-4-carbonitrile | |
85515 | |
≥98.0% (GC,T) |
16617-46-2 | |
White-Yellow | |
MFCD00005237 | |
3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole | |
C1=NNC(=C1C#N)N | |
108.104 | |
108.10 | |
Crystalline Powder |
Chemical Identifiers
16617-46-2 | |
108.104 | |
FFNKBQRKZRMYCL-UHFFFAOYSA-N | |
85515 | |
C1=NNC(=C1C#N)N |
C4H4N4 | |
MFCD00005237 | |
3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole | |
5-amino-1H-pyrazole-4-carbonitrile |
Safety and Handling
TSCA : Yes