missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-pyrazolecarbonitrile 98.0+%, TCI America™

$89.97 - $271.62

Chemical Identifiers

CAS 16617-46-2
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.104
MDL Number MFCD00005237
InChI Key FFNKBQRKZRMYCL-UHFFFAOYSA-N
Synonym 3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole
PubChem CID 85515
IUPAC Name 5-amino-1H-pyrazole-4-carbonitrile
SMILES C1=NNC(=C1C#N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A15945G
View Documents
TCI America
A15945G
5 g
Each for $89.97
 
A159425G
View Documents
TCI America
A159425G
25 g
Each for $271.62
 
Description

Description

Specifications

Chemical Identifiers

16617-46-2
108.104
FFNKBQRKZRMYCL-UHFFFAOYSA-N
85515
C1=NNC(=C1C#N)N
C4H4N4
MFCD00005237
3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole
5-amino-1H-pyrazole-4-carbonitrile

Specifications

16617-46-2
White-Yellow
MFCD00005237
3-amino-4-pyrazolecarbonitrile, 3-amino-1h-pyrazole-4-carbonitrile, 3-amino-4-cyanopyrazole, 3-aminopyrazole-4-carbonitrile, 5-amino-4-cyanopyrazole, 1h-pyrazole-4-carbonitrile, 3-amino, 5-aminopyrazole-4-carbonitrile, 3-amino-4-cyano-1h-pyrazole, 4-cyano-5-aminopyrazole, 3-amino-4-cyano pyrazole
C1=NNC(=C1C#N)N
108.104
108.10
Crystalline Powder
175°C
C4H4N4
5 g
FFNKBQRKZRMYCL-UHFFFAOYSA-N
5-amino-1H-pyrazole-4-carbonitrile
85515
≥98.0% (GC,T)
3-Amino-4-pyrazolecarbonitrile
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'