missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropyldimethylmethoxysilane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 336600050

 View more versions of this product

Catalog No. AC336600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Aminopropyldimethylmethoxysilane
31024-26-7
100.0
87°C (50.0mmHg)
Authentic
Glass bottle
1.4300 to 1.432
5g
3-aminopropyl dimethylmethoxysilane, 3-aminopropyldimethylmethoxysilane, 3-aminopropyl methoxy dimethylsilane, 3-methoxy dimethyl silyl propan-1-amine, aminopropyldimethylmethoxysilane, aminopropyl-dimethylmethoxysilane, 3-aminopropyl dimethyl methoxysilane, 1-propanamine,3-methoxydimethylsilyl, 1-propanamine, 3-methoxydimethylsilyl, 3-methoxy dimethyl silyl-1-propanamine
MCLXOMWIZZCOCA-UHFFFAOYSA-N
3-[methoxy(dimethyl)silyl]propan-1-amine
2733753
97%
97%
96.0
0.8700g/mL
38°C
96% min. (GC)
C6H17NOSi
MFCD01861689
0.87
Solubility in water: reacts
CO[Si](C)(C)CCCN
147.29
147.29

Chemical Identifiers

31024-26-7
147.29
MCLXOMWIZZCOCA-UHFFFAOYSA-N
2733753
CO[Si](C)(C)CCCN
C6H17NOSi
MFCD01861689
3-aminopropyl dimethylmethoxysilane, 3-aminopropyldimethylmethoxysilane, 3-aminopropyl methoxy dimethylsilane, 3-methoxy dimethyl silyl propan-1-amine, aminopropyldimethylmethoxysilane, aminopropyl-dimethylmethoxysilane, 3-aminopropyl dimethyl methoxysilane, 1-propanamine,3-methoxydimethylsilyl, 1-propanamine, 3-methoxydimethylsilyl, 3-methoxy dimethyl silyl-1-propanamine
3-[methoxy(dimethyl)silyl]propan-1-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only