missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropyldimethylmethoxysilane, 97%, Thermo Scientific™

Chemical Identifiers

CAS 31024-26-7
Molecular Formula C6H17NOSi
Molecular Weight (g/mol) 147.29
MDL Number MFCD01861689
InChI Key MCLXOMWIZZCOCA-UHFFFAOYSA-N
Synonym 3-aminopropyl dimethylmethoxysilane, 3-aminopropyldimethylmethoxysilane, 3-aminopropyl methoxy dimethylsilane, 3-methoxy dimethyl silyl propan-1-amine, aminopropyldimethylmethoxysilane, aminopropyl-dimethylmethoxysilane, 3-aminopropyl dimethyl methoxysilane, 1-propanamine,3-methoxydimethylsilyl, 1-propanamine, 3-methoxydimethylsilyl, 3-methoxy dimethyl silyl-1-propanamine
PubChem CID 2733753
IUPAC Name 3-[methoxy(dimethyl)silyl]propan-1-amine
SMILES CO[Si](C)(C)CCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC336600010
SDS View Documents
Thermo Scientific Chemicals
336600010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC336600050
SDS View Documents
Thermo Scientific Chemicals
336600050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

31024-26-7
147.29
MCLXOMWIZZCOCA-UHFFFAOYSA-N
2733753
CO[Si](C)(C)CCCN
C6H17NOSi
MFCD01861689
3-aminopropyl dimethylmethoxysilane, 3-aminopropyldimethylmethoxysilane, 3-aminopropyl methoxy dimethylsilane, 3-methoxy dimethyl silyl propan-1-amine, aminopropyldimethylmethoxysilane, aminopropyl-dimethylmethoxysilane, 3-aminopropyl dimethyl methoxysilane, 1-propanamine,3-methoxydimethylsilyl, 1-propanamine, 3-methoxydimethylsilyl, 3-methoxy dimethyl silyl-1-propanamine
3-[methoxy(dimethyl)silyl]propan-1-amine

Specifications

31024-26-7
100.0
87°C (50.0mmHg)
Authentic
Glass bottle
1.4300 to 1.432
1g
3-aminopropyl dimethylmethoxysilane, 3-aminopropyldimethylmethoxysilane, 3-aminopropyl methoxy dimethylsilane, 3-methoxy dimethyl silyl propan-1-amine, aminopropyldimethylmethoxysilane, aminopropyl-dimethylmethoxysilane, 3-aminopropyl dimethyl methoxysilane, 1-propanamine,3-methoxydimethylsilyl, 1-propanamine, 3-methoxydimethylsilyl, 3-methoxy dimethyl silyl-1-propanamine
MCLXOMWIZZCOCA-UHFFFAOYSA-N
3-[methoxy(dimethyl)silyl]propan-1-amine
2733753
97%
96.0
0.8700g/mL
38°C
96% min. (GC)
C6H17NOSi
MFCD01861689
0.87
Solubility in water: reacts
CO[Si](C)(C)CCCN
147.29
147.29
3-Aminopropyldimethylmethoxysilane, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only