3-(Benzyldimethylammonio)propanesulfonate, 95%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals 428070250
 View more versions of this product
Catalog No. AC428070250
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalyst
Specifications
Specifications
3-(Benzyldimethylammonio)propanesulfonate
94.0
95%
C12H19NO3S
25g
Solubility in water: 10g/L (20°C)
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
257.35
257.35
81239-45-4
100.0
Glass bottle
MFCD00225018
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
MEJASPJNLSQOAG-UHFFFAOYSA-N
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
4386830
95%
Chemical Identifiers
81239-45-4
257.35
MEJASPJNLSQOAG-UHFFFAOYSA-N
4386830
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
C12H19NO3S
MFCD00225018
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
Videos
SDS Documents