missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Benzyldimethylammonio)propanesulfonate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 81239-45-4
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.35
MDL Number MFCD00225018
InChI Key MEJASPJNLSQOAG-UHFFFAOYSA-N
Synonym ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
PubChem CID 4386830
IUPAC Name 3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428070050
SDS View Documents
Thermo Scientific Chemicals
428070050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC428070250
SDS View Documents
Thermo Scientific Chemicals
428070250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
Specifications

Chemical Identifiers

81239-45-4
257.35
MEJASPJNLSQOAG-UHFFFAOYSA-N
4386830
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
C12H19NO3S
MFCD00225018
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate

Specifications

81239-45-4
100.0
Glass bottle
MFCD00225018
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
MEJASPJNLSQOAG-UHFFFAOYSA-N
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
4386830
95%
94.0
95%
C12H19NO3S
5g
Solubility in water: 10g/L (20°C)
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
257.35
257.35
3-(Benzyldimethylammonio)propanesulfonate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only