missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-5-iodopyridine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6400703

 View more versions of this product

Catalog No. AAH6400703


Explore available promotions

Description

Description

Reactant in the synthesis chiral 4, 4?-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis chiral 4, 4′-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat, air.
Specifications

Specifications

3-Bromo-5-iodopyridine
127°C to 131°C
C5H3BrIN
1 g
Light sensitive
Slightly soluble in water.
C1=C(C=NC=C1I)Br
283.894
283.89
Solid
233770-01-9
White
MFCD03086019
108478
3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
AOOZLVWDZUPEHT-UHFFFAOYSA-N
3-bromo-5-iodopyridine
820409
95%

Chemical Identifiers

233770-01-9
283.894
AOOZLVWDZUPEHT-UHFFFAOYSA-N
3-bromo-5-iodopyridine
C5H3BrIN
MFCD03086019
820409
C1=C(C=NC=C1I)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only