Learn More
3-Bromo-5-iodopyridine, 95%, Thermo Scientific Chemicals
$242.88 - $874.74
Chemical Identifiers
CAS | 233770-01-9 |
---|---|
Molecular Formula | C5H3BrIN |
Molecular Weight (g/mol) | 283.894 |
MDL Number | MFCD03086019 |
InChI Key | AOOZLVWDZUPEHT-UHFFFAOYSA-N |
PubChem CID | 820409 |
IUPAC Name | 3-bromo-5-iodopyridine |
SMILES | C1=C(C=NC=C1I)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6400703
|
Thermo Scientific Chemicals
H6400703 |
1 g |
Each for $242.88
|
|
||||||
AAH6400706
|
Thermo Scientific Chemicals
H6400706 |
5 g |
Each for $874.74
|
|
||||||
Description
Reactant in the synthesis chiral 4, 4?-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis chiral 4, 4′-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat, air.
Chemical Identifiers
233770-01-9 | |
283.894 | |
AOOZLVWDZUPEHT-UHFFFAOYSA-N | |
3-bromo-5-iodopyridine |
C5H3BrIN | |
MFCD03086019 | |
820409 | |
C1=C(C=NC=C1I)Br |
Specifications
233770-01-9 | |
White | |
MFCD03086019 | |
108478 | |
3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787 | |
AOOZLVWDZUPEHT-UHFFFAOYSA-N | |
3-bromo-5-iodopyridine | |
820409 | |
95% | |
3-Bromo-5-iodopyridine |
127°C to 131°C | |
C5H3BrIN | |
1 g | |
Light sensitive | |
Slightly soluble in water. | |
C1=C(C=NC=C1I)Br | |
283.894 | |
283.89 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only