missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-5-methylaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6151903

 View more versions of this product

Catalog No. AAH6151903


Explore available promotions

Description

Description

3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

3-Bromo-5-methylaniline
Brown
MFCD11845553
UN2811
Slightly soluble in water.
CC1=CC(=CC(=C1)Br)N
186.052
186.05
Powder
74586-53-1
C7H8BrN
1 g
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3-bromo-5-methylaniline
3018526
98%

Chemical Identifiers

74586-53-1
186.052
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3018526
CC1=CC(=CC(=C1)Br)N
C7H8BrN
MFCD11845553
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
3-bromo-5-methylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 277-932-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only