missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methylaniline, 98%

CAS: 74586-53-1 | C7H8BrN | 186.052 g/mol

$91.79 - $349.53

Chemical Identifiers

CAS 74586-53-1
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD11845553
InChI Key YIZRPAWCIFTHNA-UHFFFAOYSA-N
Synonym 3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
PubChem CID 3018526
IUPAC Name 3-bromo-5-methylaniline
SMILES CC1=CC(=CC(=C1)Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6151903
View Documents
Thermo Scientific Chemicals
H6151903
1 g
Each for $91.79
Only null left
 
AAH6151906
View Documents
Thermo Scientific Chemicals
H6151906
5 g
Each for $349.53
Only null left
 
Description

Description

3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

74586-53-1
186.052
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3018526
CC1=CC(=CC(=C1)Br)N
C7H8BrN
MFCD11845553
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
3-bromo-5-methylaniline

Specifications

74586-53-1
C7H8BrN
1 g
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3-bromo-5-methylaniline
3018526
98%
3-Bromo-5-methylaniline
Brown
MFCD11845553
UN2811
Slightly soluble in water.
CC1=CC(=CC(=C1)Br)N
186.052
186.05
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 277-932-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only