missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzaldehyde ethylene acetal, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1961106

 View more versions of this product

Catalog No. AAL1961106

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromobenzaldehyde ethylene acetal
1.514
>110°C (230°F)
1.563
5 g
2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
C1COC(O1)C2=CC(=CC=C2)Br
229.073
229.07
17789-14-9
132°C to 133°C (8 mmHg)
C9H9BrO2
MFCD00003209
1344030
VYPYKCPWNPPBBX-UHFFFAOYSA-N
2-(3-bromophenyl)-1,3-dioxolane
87306
≥98%

Chemical Identifiers

17789-14-9
229.073
VYPYKCPWNPPBBX-UHFFFAOYSA-N
87306
C1COC(O1)C2=CC(=CC=C2)Br
C9H9BrO2
MFCD00003209
2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
2-(3-bromophenyl)-1,3-dioxolane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 241-766-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only