missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzaldehyde ethylene acetal, 98+%, Thermo Scientific Chemicals

$70.85 - $213.66

Chemical Identifiers

CAS 17789-14-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00003209
InChI Key VYPYKCPWNPPBBX-UHFFFAOYSA-N
Synonym 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
PubChem CID 87306
IUPAC Name 2-(3-bromophenyl)-1,3-dioxolane
SMILES C1COC(O1)C2=CC(=CC=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1961106
View Documents
Thermo Scientific Chemicals
L1961106
5 g N/A
 
AAL1961114
View Documents
Thermo Scientific Chemicals
L1961114
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17789-14-9
229.073
VYPYKCPWNPPBBX-UHFFFAOYSA-N
87306
C1COC(O1)C2=CC(=CC=C2)Br
C9H9BrO2
MFCD00003209
2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
2-(3-bromophenyl)-1,3-dioxolane

Specifications

17789-14-9
132°C to 133°C (8 mmHg)
C9H9BrO2
MFCD00003209
1344030
VYPYKCPWNPPBBX-UHFFFAOYSA-N
2-(3-bromophenyl)-1,3-dioxolane
87306
≥98%
1.514
>110°C (230°F)
1.563
5 g
2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
C1COC(O1)C2=CC(=CC=C2)Br
229.073
229.07
3-Bromobenzaldehyde ethylene acetal
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 241-766-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only