missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals

$65.39 - $197.66

Chemical Identifiers

CAS 823-78-9
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000176
InChI Key ZPCJPJQUVRIILS-UHFFFAOYSA-N
Synonym 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
PubChem CID 69979
IUPAC Name 1-bromo-3-(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC181780050
SDS View Documents
Thermo Scientific Chemicals
181780050
5 g Glass bottle
Each for $65.39
 
AC181780250
SDS View Documents
Thermo Scientific Chemicals
181780250
25 g Glass bottle
Each for $197.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

823-78-9
249.93
ZPCJPJQUVRIILS-UHFFFAOYSA-N
69979
C1=CC(=CC(=C1)Br)CBr
C7H6Br2
MFCD00000176
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
1-bromo-3-(bromomethyl)benzene

Specifications

823-78-9
100.0
White
>110°C
94% min. (GC)
C7H6Br2
MFCD00000176
05, 308
Solubility in water: insoluble. Other solubilities: soluble in methanol
C1=CC(=CC(=C1)Br)CBr
249.93
249.93
Adhering Crystals or Crystalline Powder
94.0
37.0°C to 42.0°C
130.0°C (12.0 mmHg)
Authentic
Glass bottle
BrC6H4CH2Br
5 g
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
ZPCJPJQUVRIILS-UHFFFAOYSA-N
1-bromo-3-(bromomethyl)benzene
69979
95%
3-Bromobenzyl bromide, 95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 212-519-1

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only