missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3365206

 View more versions of this product

Catalog No. AAH3365206


Explore available promotions

Description

Description

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

3-Bromofuro[3,2-b]pyridine
51°C to 53°C
MFCD19687763
3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
C1=CC2=C(C(=CO2)Br)N=C1
198.019
198.02
92404-63-2
C7H4BrNO
5 g
KHMAAFDMZSYVEF-UHFFFAOYSA-N
3-bromofuro[3,2-b]pyridine
641477
96%

Chemical Identifiers

92404-63-2
198.019
KHMAAFDMZSYVEF-UHFFFAOYSA-N
641477
C1=CC2=C(C(=CO2)Br)N=C1
C7H4BrNO
MFCD19687763
3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
3-bromofuro[3,2-b]pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only