missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals

$398.25 - $1324.22

Chemical Identifiers

CAS 92404-63-2
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD19687763
InChI Key KHMAAFDMZSYVEF-UHFFFAOYSA-N
Synonym 3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
PubChem CID 641477
IUPAC Name 3-bromofuro[3,2-b]pyridine
SMILES C1=CC2=C(C(=CO2)Br)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3365203
View Documents
Thermo Scientific Chemicals
H3365203
1 g
Each for $398.25
 
AAH3365206
View Documents
Thermo Scientific Chemicals
H3365206
5 g
Each for $1,324.22
 
Description

Description

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

92404-63-2
198.019
KHMAAFDMZSYVEF-UHFFFAOYSA-N
641477
C1=CC2=C(C(=CO2)Br)N=C1
C7H4BrNO
MFCD19687763
3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
3-bromofuro[3,2-b]pyridine

Specifications

92404-63-2
C7H4BrNO
1 g
KHMAAFDMZSYVEF-UHFFFAOYSA-N
3-bromofuro[3,2-b]pyridine
641477
96%
51°C to 53°C
MFCD19687763
3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
C1=CC2=C(C(=CO2)Br)N=C1
198.019
198.02
3-Bromofuro[3,2-b]pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only