missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromoquinoline, 98%, Thermo Scientific Chemicals

$205.61 - $205.61

Chemical Identifiers

CAS 5332-24-1
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
InChI Key ZGIKWINFUGEQEO-UHFFFAOYSA-N
Synonym quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
PubChem CID 21413
IUPAC Name 3-bromoquinoline
SMILES C1=CC=C2C(=C1)C=C(C=N2)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154270250
SDS View Documents
Thermo Scientific Chemicals
154270250
25 g Glass bottle
Each for $205.61
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5332-24-1
208.06
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
3-bromoquinoline
C9H6BrN
ZGIKWINFUGEQEO-UHFFFAOYSA-N
21413
C1=CC=C2C(=C1)C=C(C=N2)Br

Specifications

5332-24-1
100.0
Colorless to Yellow
274.0°C to 276.0°C
Authentic
Glass bottle
1.6630 to 1.6650
20, 363
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
C1=CC=C2C(=C1)C=C(C=N2)Br
208.06
208.06
Liquid
97.5
13.0°C to 15.0°C
1.5300g/mL
>112°C
97.5% min. (GC)
C9H6BrN
25 g
1.53
ZGIKWINFUGEQEO-UHFFFAOYSA-N
3-bromoquinoline
21413
98%
3-Bromoquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-237-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only