missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H26711MD

 View more versions of this product

Catalog No. AAH26711MD

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2,4-difluorophenylacetic acid is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

3-Chloro-2,4-difluorophenylacetic acid
107°C to 110°C
MFCD06660337
3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoro
C1=CC(=C(C(=C1CC(=O)O)F)Cl)F
206.573
206.58
886761-66-6
C8H5ClF2O2
250 mg
HENNVWWQJGUCAH-UHFFFAOYSA-N
2-(3-chloro-2,4-difluorophenyl)acetic acid
17750824
97%

Chemical Identifiers

886761-66-6
206.573
HENNVWWQJGUCAH-UHFFFAOYSA-N
17750824
C1=CC(=C(C(=C1CC(=O)O)F)Cl)F
C8H5ClF2O2
MFCD06660337
3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoro
2-(3-chloro-2,4-difluorophenyl)acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions