missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™
$149.75 - $414.55
Chemical Identifiers
CAS | 886761-66-6 |
---|---|
Molecular Formula | C8H5ClF2O2 |
Molecular Weight (g/mol) | 206.573 |
MDL Number | MFCD06660337 |
InChI Key | HENNVWWQJGUCAH-UHFFFAOYSA-N |
Synonym | 3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoro |
PubChem CID | 17750824 |
IUPAC Name | 2-(3-chloro-2,4-difluorophenyl)acetic acid |
SMILES | C1=CC(=C(C(=C1CC(=O)O)F)Cl)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH26711MD
|
Thermo Scientific Chemicals
H26711MD |
250 mg |
Each for $149.75
|
|
|||||
AAH2671103
|
Thermo Scientific Chemicals
H2671103 |
1 g |
Each for $414.55
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Chloro-2,4-difluorophenylacetic acid is used as an intermediate in organic synthesis.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
886761-66-6 | |
206.573 | |
HENNVWWQJGUCAH-UHFFFAOYSA-N | |
17750824 | |
C1=CC(=C(C(=C1CC(=O)O)F)Cl)F |
C8H5ClF2O2 | |
MFCD06660337 | |
3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoro | |
2-(3-chloro-2,4-difluorophenyl)acetic acid |
Specifications
886761-66-6 | |
C8H5ClF2O2 | |
250 mg | |
HENNVWWQJGUCAH-UHFFFAOYSA-N | |
2-(3-chloro-2,4-difluorophenyl)acetic acid | |
17750824 | |
97% |
107°C to 110°C | |
MFCD06660337 | |
3-chloro-2,4-difluorophenylacetic acid, 3-chloro-2,4-difluorophenyl acetic acid, 2-3-chloro-2,4-difluorophenyl acetic acid, benzeneacetic acid, 3-chloro-2,4-difluoro | |
C1=CC(=C(C(=C1CC(=O)O)F)Cl)F | |
206.573 | |
206.58 | |
3-Chloro-2,4-difluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures