missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 80563-86-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.09
MDL Number MFCD00052198
InChI Key ZSIYKAQPQRTBPF-UHFFFAOYSA-N
Synonym 3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
PubChem CID 522734
IUPAC Name 3-chloro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357080010
SDS View Documents
Thermo Scientific Chemicals
357080010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

80563-86-6
225.09
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
522734
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
C7H6Cl2O2S
MFCD00052198
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
3-chloro-2-methylbenzenesulfonyl chloride

Specifications

80563-86-6
100.0
97.5% min. (ex Cl) (Argentometry)
C7H6Cl2O2S
MFCD00052198
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
225.09
225.09
3-chloro-2-methylbenzenesulfonyl chloride
97.5
Authentic
Glass bottle
Cl(CH3)C6H3SO2Cl
1g
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
3-chloro-2-methylbenzenesulfonyl chloride
522734
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only