missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2-methylphenyl isocyanate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1080206

 View more versions of this product

Catalog No. AAL1080206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-2-methylphenyl isocyanate
1.247
5 g
C8H6ClNO
UN2206
3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl
CC1=C(C=CC=C1Cl)N=C=O
167.59
167.6
40397-90-8
94°C to 95°C (9 mmHg)
1.559
MFCD00037036
2691111
NGQMCUWZGVMILT-UHFFFAOYSA-N
1-chloro-3-isocyanato-2-methylbenzene
2735770
99%

Chemical Identifiers

40397-90-8
167.59
NGQMCUWZGVMILT-UHFFFAOYSA-N
2735770
CC1=C(C=CC=C1Cl)N=C=O
C8H6ClNO
MFCD00037036
3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl
1-chloro-3-isocyanato-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H319-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only