Learn More
3-Chloro-2-methylphenyl isocyanate, 99%, Thermo Scientific Chemicals
$54.67 - $163.53
Chemical Identifiers
CAS | 40397-90-8 |
---|---|
Molecular Formula | C8H6ClNO |
Molecular Weight (g/mol) | 167.59 |
MDL Number | MFCD00037036 |
InChI Key | NGQMCUWZGVMILT-UHFFFAOYSA-N |
Synonym | 3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl |
PubChem CID | 2735770 |
IUPAC Name | 1-chloro-3-isocyanato-2-methylbenzene |
SMILES | CC1=C(C=CC=C1Cl)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1080203
|
Thermo Scientific Chemicals
L1080203 |
1 g |
Each for $54.67
|
|
|||||
AAL1080206
|
Thermo Scientific Chemicals
L1080206 |
5 g |
Each for $163.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40397-90-8 | |
167.59 | |
NGQMCUWZGVMILT-UHFFFAOYSA-N | |
2735770 | |
CC1=C(C=CC=C1Cl)N=C=O |
C8H6ClNO | |
MFCD00037036 | |
3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl | |
1-chloro-3-isocyanato-2-methylbenzene |
Specifications
40397-90-8 | |
94°C to 95°C (9 mmHg) | |
1.559 | |
MFCD00037036 | |
2691111 | |
NGQMCUWZGVMILT-UHFFFAOYSA-N | |
1-chloro-3-isocyanato-2-methylbenzene | |
2735770 | |
99% |
1.247 | |
1 g | |
C8H6ClNO | |
UN2206 | |
3-chloro-2-methylphenyl isocyanate, 3-chloro-2-methylphenylisocyanate, 1-chloro-3-isocyanato-2-methyl-benzene, 3-chloro-2-methylbenzenisocyanate, acmc-1aqe4, 2-chloro-6-isocyanatotoluene, benzene,1-chloro-3-isocyanato-2-methyl, 1-chloranyl-3-isocyanato-2-methyl-benzene, benzene, 1-chloro-3-isocyanato-2-methyl | |
CC1=C(C=CC=C1Cl)N=C=O | |
167.59 | |
167.6 | |
3-Chloro-2-methylphenyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H319-H330-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only