missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 430620100

 View more versions of this product

Catalog No. AC430620100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-chloro-4-methylphenylboronic acid
96.0
Authentic
Glass bottle
MFCD04039010
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
B(C1=CC(=C(C=C1)C)Cl)(O)O
170.4
170.4
175883-63-3
100.0
96% min. (HPLC)
C7H8BClO2
10g
YTJUYWRCAZWVSX-UHFFFAOYSA-N
(3-chloro-4-methylphenyl)boronic acid
3854610
97%

Chemical Identifiers

175883-63-3
170.4
YTJUYWRCAZWVSX-UHFFFAOYSA-N
3854610
B(C1=CC(=C(C=C1)C)Cl)(O)O
C7H8BClO2
MFCD04039010
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
(3-chloro-4-methylphenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only