missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 175883-63-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.4
MDL Number MFCD04039010
InChI Key YTJUYWRCAZWVSX-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
PubChem CID 3854610
IUPAC Name (3-chloro-4-methylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC430620010
SDS View Documents
Thermo Scientific Chemicals
430620010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC430620100
SDS View Documents
Thermo Scientific Chemicals
430620100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

175883-63-3
170.4
YTJUYWRCAZWVSX-UHFFFAOYSA-N
3854610
B(C1=CC(=C(C=C1)C)Cl)(O)O
C7H8BClO2
MFCD04039010
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
(3-chloro-4-methylphenyl)boronic acid

Specifications

175883-63-3
100.0
96% min. (HPLC)
C7H8BClO2
1g
YTJUYWRCAZWVSX-UHFFFAOYSA-N
(3-chloro-4-methylphenyl)boronic acid
3854610
97%
96.0
Authentic
Glass bottle
MFCD04039010
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
B(C1=CC(=C(C=C1)C)Cl)(O)O
170.4
170.4
3-chloro-4-methylphenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only