missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2619506

 View more versions of this product

Catalog No. AAH2619506


Explore available promotions

Description

Description

3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

3-Chloro-5-fluorophenol
C6H4ClFO
MFCD00042212
3265
Immiscible with water.
C1=C(C=C(C=C1F)Cl)O
146.545
146.55
202982-70-5
1.537
5 g
3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci
ZMFBYTAAMHWQHD-UHFFFAOYSA-N
3-chloro-5-fluorophenol
2773713
98%

Chemical Identifiers

202982-70-5
146.545
ZMFBYTAAMHWQHD-UHFFFAOYSA-N
2773713
C1=C(C=C(C=C1F)Cl)O
C6H4ClFO
MFCD00042212
3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci
3-chloro-5-fluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only