Learn More
3-Chloro-5-fluorophenol, 98%
CAS: 202982-70-5 | C6H4ClFO | 146.545 g/mol
$160.12 - $466.04
Chemical Identifiers
| CAS | 202982-70-5 |
|---|---|
| Molecular Formula | C6H4ClFO |
| Molecular Weight (g/mol) | 146.545 |
| MDL Number | MFCD00042212 |
| InChI Key | ZMFBYTAAMHWQHD-UHFFFAOYSA-N |
| Synonym | 3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci |
| PubChem CID | 2773713 |
| IUPAC Name | 3-chloro-5-fluorophenol |
| SMILES | C1=C(C=C(C=C1F)Cl)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2619503
|
Thermo Scientific Chemicals
H2619503 |
1 g |
Each for $160.12
|
|
|||||
|
AAH2619506
|
Thermo Scientific Chemicals
H2619506 |
5 g |
Each for $466.04
|
|
|||||
Description
3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 202982-70-5 | |
| 146.545 | |
| ZMFBYTAAMHWQHD-UHFFFAOYSA-N | |
| 2773713 | |
| C1=C(C=C(C=C1F)Cl)O |
| C6H4ClFO | |
| MFCD00042212 | |
| 3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci | |
| 3-chloro-5-fluorophenol |
Specifications
| 202982-70-5 | |
| 1.537 | |
| 1 g | |
| 3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci | |
| ZMFBYTAAMHWQHD-UHFFFAOYSA-N | |
| 3-chloro-5-fluorophenol | |
| 2773713 | |
| 98% |
| C6H4ClFO | |
| MFCD00042212 | |
| UN3265 | |
| Immiscible with water. | |
| C1=C(C=C(C=C1F)Cl)O | |
| 146.545 | |
| 146.55 | |
| 3-Chloro-5-fluorophenol |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only