Learn More
3-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1325414
Description
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
Solubility
It is soluble in alcohol, ether, and acetone. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong bases, Strong oxidizing agents, Strong reducing agents.
Specifications
3-Chlorobenzaldehyde | |
16°C to 18°C | |
213°C to 214°C | |
Characteristic | |
1.564 | |
25 g | |
Air Sensitive | |
It is soluble in alcohol,ether,and acetone. Insoluble in water. | |
C1=CC(=CC(=C1)Cl)C=O | |
140.566 | |
140.57 |
587-04-2 | |
1.239 | |
88°C (190°F) | |
C7H5ClO | |
MFCD00003350 | |
507098 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
3-chlorobenzaldehyde | |
11477 | |
97% |
Chemical Identifiers
587-04-2 | |
140.566 | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
11477 | |
C1=CC(=CC(=C1)Cl)C=O |
C7H5ClO | |
MFCD00003350 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
3-chlorobenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-596-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only