Learn More
3-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals
$103.73 - $1232.67
Chemical Identifiers
CAS | 587-04-2 |
---|---|
Molecular Formula | C7H5ClO |
Molecular Weight (g/mol) | 140.566 |
MDL Number | MFCD00003350 |
InChI Key | SRWILAKSARHZPR-UHFFFAOYSA-N |
Synonym | m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde |
PubChem CID | 11477 |
IUPAC Name | 3-chlorobenzaldehyde |
SMILES | C1=CC(=CC(=C1)Cl)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1325414
|
Thermo Scientific Chemicals
A1325414 |
25 g |
Each for $103.73
|
|
||||||
AAA1325422
|
Thermo Scientific Chemicals
A1325422 |
100 g |
Each for $323.23
|
|
||||||
AAA1325436
|
Thermo Scientific Chemicals
A1325436 |
500 g |
Each for $1,232.67
|
|
||||||
Description
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
Solubility
It is soluble in alcohol, ether, and acetone. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong bases, Strong oxidizing agents, Strong reducing agents.
Chemical Identifiers
587-04-2 | |
140.566 | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
11477 | |
C1=CC(=CC(=C1)Cl)C=O |
C7H5ClO | |
MFCD00003350 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
3-chlorobenzaldehyde |
Specifications
587-04-2 | |
1.239 | |
88°C (190°F) | |
C7H5ClO | |
MFCD00003350 | |
507098 | |
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
SRWILAKSARHZPR-UHFFFAOYSA-N | |
3-chlorobenzaldehyde | |
11477 | |
97% |
16°C to 18°C | |
213°C to 214°C | |
Characteristic | |
1.564 | |
25 g | |
Air Sensitive | |
It is soluble in alcohol,ether,and acetone. Insoluble in water. | |
C1=CC(=CC(=C1)Cl)C=O | |
140.566 | |
140.57 | |
3-Chlorobenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-596-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only