missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1087622

 View more versions of this product

Catalog No. AAA1087622


Explore available promotions

Description

Description

A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

Solubility
Slioghtly soluble in water(0.45 g/L ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-Chlorobenzoic acid
154°C to 160°C
Odorless
MFCD00002491
907218
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
LULAYUGMBFYYEX-UHFFFAOYSA-N
3-chlorobenzoic acid
447
156.57
535-80-8
274°C to 276°C
C7H5ClO2
100 g
14,2123
Slioghtly soluble in water(0.45g/L ).
C1=CC(=CC(=C1)Cl)C(=O)O
156.565
CHEBI:49410
99%

Chemical Identifiers

535-80-8
156.565
LULAYUGMBFYYEX-UHFFFAOYSA-N
447
3-chlorobenzoic acid
C7H5ClO2
MFCD00002491
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
CHEBI:49410
C1=CC(=CC(=C1)Cl)C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 208-618-4

RTECSNumber : DG4975990

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only