Learn More
3-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
$55.88 - $418.19
Chemical Identifiers
CAS | 535-80-8 |
---|---|
Molecular Formula | C7H5ClO2 |
Molecular Weight (g/mol) | 156.565 |
MDL Number | MFCD00002491 |
InChI Key | LULAYUGMBFYYEX-UHFFFAOYSA-N |
Synonym | m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992 |
PubChem CID | 447 |
ChEBI | CHEBI:49410 |
IUPAC Name | 3-chlorobenzoic acid |
SMILES | C1=CC(=CC(=C1)Cl)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1087614
|
Thermo Scientific Chemicals
A1087614 |
25 g |
Each for $55.88
|
|
|||||
AAA1087622
|
Thermo Scientific Chemicals
A1087622 |
100 g |
Each for $142.44
|
|
|||||
AAA1087636
|
Thermo Scientific Chemicals
A1087636 |
500 g |
Each for $418.19
|
|
|||||
Description
A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.
Solubility
Slioghtly soluble in water(0.45 g/L ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
535-80-8 | |
156.565 | |
LULAYUGMBFYYEX-UHFFFAOYSA-N | |
447 | |
3-chlorobenzoic acid |
C7H5ClO2 | |
MFCD00002491 | |
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992 | |
CHEBI:49410 | |
C1=CC(=CC(=C1)Cl)C(=O)O |
Specifications
535-80-8 | |
274°C to 276°C | |
C7H5ClO2 | |
25 g | |
14,2123 | |
Slioghtly soluble in water(0.45g/L ). | |
C1=CC(=CC(=C1)Cl)C(=O)O | |
156.565 | |
CHEBI:49410 | |
99% |
154°C to 160°C | |
Odorless | |
MFCD00002491 | |
907218 | |
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992 | |
LULAYUGMBFYYEX-UHFFFAOYSA-N | |
3-chlorobenzoic acid | |
447 | |
156.57 | |
3-Chlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 208-618-4
RTECSNumber : DG4975990
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only