missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclohexylpropionic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2361318

 View more versions of this product

Catalog No. AAB2361318


Explore available promotions

Description

Description

3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

Solubility
Soluble in water 10g/ L.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases.
Specifications

Specifications

3-Cyclohexylpropionic acid
14°C to 17°C
276°C
Unpleasant
1.4636
50 g
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
HJZLEGIHUQOJBA-UHFFFAOYSA-N
3-cyclohexylpropanoic acid
69702
≥98%
701-97-3
0.99
130°C (266°F)
C9H16O2
MFCD00001527
2042799
Soluble in water 10g/ L.
OC(=O)CCC1CCCCC1
156.23
156.23

Chemical Identifiers

701-97-3
156.23
HJZLEGIHUQOJBA-UHFFFAOYSA-N
69702
OC(=O)CCC1CCCCC1
C9H16O2
MFCD00001527
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
3-cyclohexylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 211-861-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only