Learn More
3-Cyclohexylpropionic acid, 98+%, Thermo Scientific Chemicals
$107.70 - $432.19
Chemical Identifiers
CAS | 701-97-3 |
---|---|
Molecular Formula | C9H16O2 |
Molecular Weight (g/mol) | 156.23 |
MDL Number | MFCD00001527 |
InChI Key | HJZLEGIHUQOJBA-UHFFFAOYSA-N |
Synonym | cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid |
PubChem CID | 69702 |
IUPAC Name | 3-cyclohexylpropanoic acid |
SMILES | OC(=O)CCC1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2361318
|
Thermo Scientific Chemicals
B2361318 |
50 g |
Each for $107.70
|
|
|||||
AAB2361330
|
Thermo Scientific Chemicals
B2361330 |
250 g |
Each for $432.19
|
|
|||||
Description
3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.
Solubility
Soluble in water 10g/ L.
Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases.
Chemical Identifiers
701-97-3 | |
156.23 | |
HJZLEGIHUQOJBA-UHFFFAOYSA-N | |
69702 | |
OC(=O)CCC1CCCCC1 |
C9H16O2 | |
MFCD00001527 | |
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid | |
3-cyclohexylpropanoic acid |
Specifications
701-97-3 | |
0.99 | |
130°C (266°F) | |
C9H16O2 | |
MFCD00001527 | |
2042799 | |
Soluble in water 10g/ L. | |
OC(=O)CCC1CCCCC1 | |
156.23 | |
156.23 | |
3-Cyclohexylpropionic acid |
14°C to 17°C | |
276°C | |
Unpleasant | |
1.4636 | |
50 g | |
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid | |
HJZLEGIHUQOJBA-UHFFFAOYSA-N | |
3-cyclohexylpropanoic acid | |
69702 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 211-861-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only