missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclopentyloxy-4-methoxybenzyl alcohol, 97%, Thermo Scientific™

$79.67 - $1157.95

Chemical Identifiers

CAS 133332-49-7
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD03844627
InChI Key JIRHAGAOHOYLNO-UHFFFAOYSA-N
Synonym 3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol
PubChem CID 11138706
IUPAC Name (3-cyclopentyloxy-4-methoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC2CCCC2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62441MD
View Documents Promotion Details
Thermo Scientific Chemicals
H62441MD
250 mg
Each for $79.67
 
AAH6244103
View Documents Promotion Details
Thermo Scientific Chemicals
H6244103
1 g
Each for $243.56
 
AAH6244106
View Documents Promotion Details
Thermo Scientific Chemicals
H6244106
5 g
Each for $1,157.95
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

133332-49-7
222.284
JIRHAGAOHOYLNO-UHFFFAOYSA-N
11138706
COC1=C(C=C(C=C1)CO)OC2CCCC2
C13H18O3
MFCD03844627
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol
(3-cyclopentyloxy-4-methoxyphenyl)methanol

Specifications

133332-49-7
MFCD03844627
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol
COC1=C(C=C(C=C1)CO)OC2CCCC2
222.284
222.28
3-Cyclopentyloxy-4-methoxybenzyl alcohol
C13H18O3
250 mg
JIRHAGAOHOYLNO-UHFFFAOYSA-N
(3-cyclopentyloxy-4-methoxyphenyl)methanol
11138706
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'