missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119550500

 View more versions of this product

Catalog No. AC119550500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluorophenol
97.5
10.0°C
1.2400g/mL
71°C
97.5% min. (GC)
C6H5FO
FC6H4OH
50 mL
1.24
Solubility in water: insoluble
C1=CC(=CC(=C1)F)O
112.1
112.1
Liquid
372-20-3
100.0
Brown-Yellow to Colorless
178.0°C
Authentic
Glass bottle
1.5125 to 1.5155
MFCD00002254
06, I, 97
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
SJTBRFHBXDZMPS-UHFFFAOYSA-N
3-fluorophenol
9743
98%

Chemical Identifiers

372-20-3
112.1
SJTBRFHBXDZMPS-UHFFFAOYSA-N
9743
C1=CC(=CC(=C1)F)O
C6H5FO
MFCD00002254
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
3-fluorophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 206-748-6

Recommended Storage : this product darkens during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only