missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorophenol, 98%, Thermo Scientific Chemicals

$120.93 - $310.55

Chemical Identifiers

CAS 372-20-3
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.1
MDL Number MFCD00002254
InChI Key SJTBRFHBXDZMPS-UHFFFAOYSA-N
Synonym m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
PubChem CID 9743
IUPAC Name 3-fluorophenol
SMILES C1=CC(=CC(=C1)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119550100
SDS View Documents
Thermo Scientific Chemicals
119550100
10 mL Glass bottle
Each for $120.93
 
AC119550500
SDS View Documents
Thermo Scientific Chemicals
119550500
50 mL Glass bottle
Each for $310.55
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

372-20-3
112.1
SJTBRFHBXDZMPS-UHFFFAOYSA-N
9743
C1=CC(=CC(=C1)F)O
C6H5FO
MFCD00002254
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
3-fluorophenol

Specifications

372-20-3
100.0
Brown-Yellow to Colorless
178.0°C
Authentic
Glass bottle
1.5125 to 1.5155
MFCD00002254
06, I, 97
m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
SJTBRFHBXDZMPS-UHFFFAOYSA-N
3-fluorophenol
9743
98%
3-Fluorophenol
97.5
10.0°C
1.2400g/mL
71°C
97.5% min. (GC)
C6H5FO
FC6H4OH
10 mL
1.24
Solubility in water: insoluble
C1=CC(=CC(=C1)F)O
112.1
112.1
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 206-748-6

Recommended Storage : this product darkens during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only