missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Furaldehyde, 98%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 393260050

 View more versions of this product

Catalog No. AC393260050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Furaldehyde
97.5
Yellow to Brown
145.0°C
Authentic
Glass bottle
1.4930 to 1.4950 (20°C, 589nm)
1.15
AZVSIHIBYRHSLB-UHFFFAOYSA-N
furan-3-carbaldehyde
10351
96.08
Liquid
498-60-2
100.0
1.1500g/mL
48°C
97.5% min. (GC)
C5H4O2
5 g
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
C1=COC=C1C=O
96.08
CHEBI:87609
98%

Chemical Identifiers

498-60-2
96.08
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
CHEBI:87609
C1=COC=C1C=O
C5H4O2
AZVSIHIBYRHSLB-UHFFFAOYSA-N
10351
furan-3-carbaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only