missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hexanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2021014

 View more versions of this product

Catalog No. AAB2021014

Explore available promotions

Description

Description

It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Hexanol
0.81
42°C (108°F)
C6H14O
MFCD00004582
UN2282
3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
ZOCHHNOQQHDWHG-UHFFFAOYSA-N
hexan-3-ol
12178
98%
623-37-0
134°C to 136°C
Characteristic
1.415
25 g
1718964
Soluble in alcohol.
CCCC(CC)O
102.177
102.18

Chemical Identifiers

623-37-0
102.177
ZOCHHNOQQHDWHG-UHFFFAOYSA-N
12178
CCCC(CC)O
C6H14O
MFCD00004582
3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
hexan-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c

H226-H302-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS

EINECSNumber : 210-790-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only