missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hexanol, 98%, Thermo Scientific Chemicals

$143.36 - $441.93

Chemical Identifiers

CAS 623-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004582
InChI Key ZOCHHNOQQHDWHG-UHFFFAOYSA-N
Synonym 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
PubChem CID 12178
IUPAC Name hexan-3-ol
SMILES CCCC(CC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2021014
View Documents
Thermo Scientific Chemicals
B2021014
25 g
Each for $143.36
 
AAB2021022
View Documents
Thermo Scientific Chemicals
B2021022
100 g
Each for $441.93
 
Description

Description

It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

623-37-0
102.177
ZOCHHNOQQHDWHG-UHFFFAOYSA-N
12178
CCCC(CC)O
C6H14O
MFCD00004582
3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
hexan-3-ol

Specifications

623-37-0
134°C to 136°C
Characteristic
1.415
25 g
1718964
Soluble in alcohol.
CCCC(CC)O
102.177
102.18
3-Hexanol
0.81
42°C (108°F)
C6H14O
MFCD00004582
UN2282
3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
ZOCHHNOQQHDWHG-UHFFFAOYSA-N
hexan-3-ol
12178
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c

H226-H302-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS

EINECSNumber : 210-790-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only