missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hexylthiophene, 98%, Thermo Scientific Chemicals

$187.54 - $733.76

Chemical Identifiers

CAS 1693-86-3
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.3
MDL Number MFCD00143181
InChI Key JEDHEMYZURJGRQ-UHFFFAOYSA-N
Synonym 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
PubChem CID 566849
IUPAC Name 3-hexylthiophene
SMILES CCCCCCC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367380050
SDS View Documents
Thermo Scientific Chemicals
367380050
5 g Glass bottle
Each for $187.54
 
AC367380250
SDS View Documents
Thermo Scientific Chemicals
367380250
25 g Glass bottle
Each for $733.76
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1693-86-3
168.3
JEDHEMYZURJGRQ-UHFFFAOYSA-N
566849
CCCCCCC1=CSC=C1
C10H16S
MFCD00143181
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
3-hexylthiophene

Specifications

1693-86-3
100.0
0.9360g/mL
37°C
97.5% min. (GC)
C10H16S
MFCD00143181
0.936
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-hexylthiophene
566849
98%
3-Hexylthiophene
97.5
Colorless to Yellow or Red
65.0°C (0.4 mmHg)
Authentic
Glass bottle
1.4945 to 1.4975
5 g
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
CCCCCCC1=CSC=C1
168.3
168.3
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.
Harmful in contact with skin.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for brea

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only