missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2568214

 View more versions of this product

Catalog No. AAB2568214


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-2-nitrobenzoic acid
219°C to 223°C
MFCD00007180
3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640
CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O
181.15
181.15
5437-38-7
C8H7NO4
25 g
DGDAVTPQCQXLGU-UHFFFAOYSA-N
3-methyl-2-nitrobenzoic acid
21575
98%

Chemical Identifiers

5437-38-7
181.15
DGDAVTPQCQXLGU-UHFFFAOYSA-N
21575
CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O
C8H7NO4
MFCD00007180
3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640
3-methyl-2-nitrobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-610-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only