missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-2-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
$48.22 - $136.05
Chemical Identifiers
CAS | 5437-38-7 |
---|---|
Molecular Formula | C8H7NO4 |
Molecular Weight (g/mol) | 181.15 |
MDL Number | MFCD00007180 |
InChI Key | DGDAVTPQCQXLGU-UHFFFAOYSA-N |
Synonym | 3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640 |
PubChem CID | 21575 |
IUPAC Name | 3-methyl-2-nitrobenzoic acid |
SMILES | CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2568214
|
Thermo Scientific Chemicals
B2568214 |
25 g |
Each for $48.22
|
|
|||||
AAB2568222
|
Thermo Scientific Chemicals
B2568222 |
100 g |
Each for $136.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5437-38-7 | |
181.15 | |
DGDAVTPQCQXLGU-UHFFFAOYSA-N | |
21575 | |
CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O |
C8H7NO4 | |
MFCD00007180 | |
3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640 | |
3-methyl-2-nitrobenzoic acid |
Specifications
5437-38-7 | |
C8H7NO4 | |
25 g | |
DGDAVTPQCQXLGU-UHFFFAOYSA-N | |
3-methyl-2-nitrobenzoic acid | |
21575 | |
98% |
219°C to 223°C | |
MFCD00007180 | |
3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640 | |
CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O | |
181.15 | |
181.15 | |
3-Methyl-2-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-610-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only