Learn More
3-Methyl-3-methoxybutanol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 259752500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methyl-3-methoxybutanol | |
56539-66-3 | |
100.0 | |
Colorless | |
174.0°C | |
Authentic | |
Glass bottle | |
1.4260 to 1.4280 | |
MFCD00044771 | |
0.926 | |
Solubility in water: freely soluble | |
CC(C)(CCO)OC | |
118.17 | |
7.35 mPa.s (20°C) | |
99% |
99% | |
98.5 | |
-50.0°C | |
0.9260g/mL | |
71°C | |
98.5% min. (GC) | |
C6H14O2 | |
(CH3)2C(OCH3)CH2CH2OH | |
250 mL | |
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389 | |
MFKRHJVUCZRDTF-UHFFFAOYSA-N | |
3-methoxy-3-methylbutan-1-ol | |
62118 | |
118.17 | |
Liquid |
Chemical Identifiers
56539-66-3 | |
MFKRHJVUCZRDTF-UHFFFAOYSA-N | |
62118 | |
CC(C)(CCO)OC |
MFCD00044771 | |
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389 | |
3-methoxy-3-methylbutan-1-ol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear face protection.
GHS Signal Word: Warning
EINECSNumber : 260-252-4
TSCA : TSCA
RUO – Research Use Only