missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-3-methoxybutanol, 99%, Thermo Scientific Chemicals

$84.32 - $84.32

Chemical Identifiers

CAS 56539-66-3
MDL Number MFCD00044771
InChI Key MFKRHJVUCZRDTF-UHFFFAOYSA-N
Synonym 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
PubChem CID 62118
IUPAC Name 3-methoxy-3-methylbutan-1-ol
SMILES CC(C)(CCO)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259752500
SDS View Documents
Thermo Scientific Chemicals
259752500
250 mL Glass bottle
Each for $84.32
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56539-66-3
MFKRHJVUCZRDTF-UHFFFAOYSA-N
62118
CC(C)(CCO)OC
MFCD00044771
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
3-methoxy-3-methylbutan-1-ol

Specifications

56539-66-3
100.0
Colorless
174.0°C
Authentic
Glass bottle
1.4260 to 1.4280
MFCD00044771
0.926
Solubility in water: freely soluble
CC(C)(CCO)OC
118.17
7.35 mPa.s (20°C)
99%
3-Methyl-3-methoxybutanol, 99%
98.5
-50.0°C
0.9260g/mL
71°C
98.5% min. (GC)
C6H14O2
(CH3)2C(OCH3)CH2CH2OH
250 mL
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
MFKRHJVUCZRDTF-UHFFFAOYSA-N
3-methoxy-3-methylbutan-1-ol
62118
118.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear face protection.

GHS Signal Word: Warning

EINECSNumber : 260-252-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only