missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylamino-1,2-propanediol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1265322

 View more versions of this product

Catalog No. AAL1265322


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methylamino-1,2-propanediol
1.093
152°C (305°F)
1.4745
100 g
1733341
WOMTYMDHLQTCHY-UHFFFAOYSA-N
3-(methylamino)propane-1,2-diol
2734122
99%
40137-22-2
239°C to 241°C
C4H11NO2
MFCD00082732
UN2735
3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
CNCC(CO)O
105.137
105.14

Chemical Identifiers

40137-22-2
105.137
WOMTYMDHLQTCHY-UHFFFAOYSA-N
2734122
CNCC(CO)O
C4H11NO2
MFCD00082732
3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
3-(methylamino)propane-1,2-diol
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 254-809-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only