missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylamino-1,2-propanediol, 99%, Thermo Scientific Chemicals

$81.11 - $233.07

Chemical Identifiers

CAS 40137-22-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00082732
InChI Key WOMTYMDHLQTCHY-UHFFFAOYSA-N
Synonym 3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
PubChem CID 2734122
IUPAC Name 3-(methylamino)propane-1,2-diol
SMILES CNCC(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1265314
View Documents Promotion Details
Thermo Scientific Chemicals
L1265314
25 g
Each for $81.11
 
AAL1265322
View Documents Promotion Details
Thermo Scientific Chemicals
L1265322
100 g
Each for $233.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40137-22-2
105.137
WOMTYMDHLQTCHY-UHFFFAOYSA-N
2734122
CNCC(CO)O
C4H11NO2
MFCD00082732
3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
3-(methylamino)propane-1,2-diol

Specifications

40137-22-2
239°C to 241°C
C4H11NO2
MFCD00082732
UN2735
3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
CNCC(CO)O
105.137
105.14
3-Methylamino-1,2-propanediol
1.093
152°C (305°F)
1.4745
25 g
1733341
WOMTYMDHLQTCHY-UHFFFAOYSA-N
3-(methylamino)propane-1,2-diol
2734122
99%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 254-809-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only