missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33809DA

 View more versions of this product

Catalog No. CC33809DA


Explore available promotions

Description

Description

Specifications

Specifications

(3-Morpholinophenyl)methanol
96.5
97%
C11H15NO2
1 g
MXBVALXTJZMIJB-UHFFFAOYSA-N
(3-morpholin-4-ylphenyl)methanol
7162074
97%
145127-38-4
100.0
Amber Glass Bottle
MFCD07772812
3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
OCC1=CC(=CC=C1)N1CCOCC1
193.25
193.24

Chemical Identifiers

145127-38-4
193.25
MXBVALXTJZMIJB-UHFFFAOYSA-N
(3-morpholin-4-ylphenyl)methanol
C11H15NO2
MFCD07772812
7162074
OCC1=CC(=CC=C1)N1CCOCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions