missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

$222.26 - $222.26

Chemical Identifiers

CAS 145127-38-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD07772812
InChI Key MXBVALXTJZMIJB-UHFFFAOYSA-N
PubChem CID 7162074
IUPAC Name (3-morpholin-4-ylphenyl)methanol
SMILES OCC1=CC(=CC=C1)N1CCOCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC33809DA
SDS View Documents
Thermo Scientific Chemicals
CC33809DA
1 g Amber Glass Bottle
Each for $222.26
 
Description

Description

Specifications

Chemical Identifiers

145127-38-4
193.25
MXBVALXTJZMIJB-UHFFFAOYSA-N
(3-morpholin-4-ylphenyl)methanol
C11H15NO2
MFCD07772812
7162074
OCC1=CC(=CC=C1)N1CCOCC1

Specifications

145127-38-4
100.0
Amber Glass Bottle
MFCD07772812
3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
OCC1=CC(=CC=C1)N1CCOCC1
193.25
193.24
(3-Morpholinophenyl)methanol
96.5
97%
C11H15NO2
1 g
MXBVALXTJZMIJB-UHFFFAOYSA-N
(3-morpholin-4-ylphenyl)methanol
7162074
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'