Learn More
3-Nitrobenzyl alcohol, 98%, Thermo Scientific Chemicals
$76.93 - $219.96
Chemical Identifiers
CAS | 619-25-0 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.137 |
MDL Number | MFCD00007273 |
InChI Key | CWNPOQFCIIFQDM-UHFFFAOYSA-N |
Synonym | 3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol |
PubChem CID | 69267 |
IUPAC Name | (3-nitrophenyl)methanol |
SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1504014
|
Thermo Scientific Chemicals
A1504014 |
25 g |
Each for $76.93
|
|
|||||
AAA1504022
|
Thermo Scientific Chemicals
A1504022 |
100 g |
Each for $219.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
619-25-0 | |
153.137 | |
CWNPOQFCIIFQDM-UHFFFAOYSA-N | |
69267 | |
C1=CC(=CC(=C1)[N+](=O)[O-])CO |
C7H7NO3 | |
MFCD00007273 | |
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol | |
(3-nitrophenyl)methanol |
Specifications
619-25-0 | |
1.3 | |
>110°C (230°F) | |
1.5755 | |
25 g | |
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol | |
C1=CC(=CC(=C1)[N+](=O)[O-])CO | |
153.137 | |
153.14 | |
3-Nitrobenzyl alcohol |
29°C to 33°C | |
175°C to 180°C (3 mmHg) | |
C7H7NO3 | |
MFCD00007273 | |
2044769 | |
CWNPOQFCIIFQDM-UHFFFAOYSA-N | |
(3-nitrophenyl)methanol | |
69267 | |
98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 210-588-2
RTECSNumber : DP0657000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only