missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 344700010

 View more versions of this product

Catalog No. AC344700010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nitrophenylboronic acid
13331-27-6
100.0
Brown to Yellow
97%
C6H6BNO4
MFCD00007193
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
(3-nitrophenyl)boronic acid
1677
97%
97%
96.0
289.0°C to 291.0°C
Authentic
Glass bottle
NO2C6H4B(OH)2
1 g
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
166.93
166.93
Powder

Chemical Identifiers

13331-27-6
166.93
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
1677
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
C6H6BNO4
MFCD00007193
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
(3-nitrophenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

RTECSNumber : CY8980000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only