missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

$39.72 - $91.93

Chemical Identifiers

CAS 13331-27-6
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00007193
InChI Key ZNRGSYUVFVNSAW-UHFFFAOYSA-N
Synonym 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
PubChem CID 1677
IUPAC Name (3-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC344700010
SDS View Documents
Thermo Scientific Chemicals
344700010
1 g Glass bottle
Each for $39.72
 
AC344700050
SDS View Documents
Thermo Scientific Chemicals
344700050
5 g Glass bottle
Each for $91.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13331-27-6
166.93
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
1677
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
C6H6BNO4
MFCD00007193
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
(3-nitrophenyl)boronic acid

Specifications

13331-27-6
100.0
Brown to Yellow
97%
C6H6BNO4
MFCD00007193
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
(3-nitrophenyl)boronic acid
1677
97%
3-Nitrophenylboronic acid, 97%
96.0
289.0°C to 291.0°C
Authentic
Glass bottle
NO2C6H4B(OH)2
1 g
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
166.93
166.93
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

RTECSNumber : CY8980000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only