Learn More
3-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 344700050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitrophenylboronic acid | |
13331-27-6 | |
100.0 | |
Brown to Yellow | |
97% | |
C6H6BNO4 | |
MFCD00007193 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
(3-nitrophenyl)boronic acid | |
1677 | |
97% |
97% | |
96.0 | |
289.0°C to 291.0°C | |
Authentic | |
Glass bottle | |
NO2C6H4B(OH)2 | |
5 g | |
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O | |
166.93 | |
166.93 | |
Powder |
Chemical Identifiers
13331-27-6 | |
166.93 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
1677 | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
C6H6BNO4 | |
MFCD00007193 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
(3-nitrophenyl)boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RTECSNumber : CY8980000
TSCA : TSCA
RUO – Research Use Only